Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC C-telopeptide TFA | 868.89 (free base) | 25 MG
SDP

Catalog No. 5000426475
Encompass_Preferred

C-telopeptide TFA is a cross-linked peptide of type I collagen. It is released during bone resorption and has been correlated with bone mineral density (BMD). This product is intended for research use only.

  • Cross-linked peptide of type I collagen
  • Released during bone resorption
  • Correlated with bone mineral density (BMD)
  • Sequence: Glu-Lys-Ala-His-Asp-Gly-Gly-Arg
  • Target: MMP
  • Pathway: Metabolic Enzyme/Protease

Catalog No. 50-004-26475 Supplier Medchemexpress LLC Supplier No. 25MGHYP0284A
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

C-telopeptide TFA is a cross-linked peptide of type I collagen. It is released during bone resorption and has been correlated with bone mineral density (BMD). This product is intended for research use only.

  • Cross-linked peptide of type I collagen
  • Released during bone resorption
  • Correlated with bone mineral density (BMD)
  • Sequence: Glu-Lys-Ala-His-Asp-Gly-Gly-Arg
  • Target: MMP
  • Pathway: Metabolic Enzyme/Protease

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.