Learn More
Enzo Life Sciences C. trachomatis native antigen (1ml)
SDP
Supplier:  Enzo Life Sciences ALX6301241000
Encompass_Preferred
Alternative name: Chlamydia trachomatis native antigen. Formulation: Liquid. In PBS containing 0.05% Triton X-100. Source: From Chlamydia trachomatis, serotype LGV2 strain 434. Inactivated by heat treatment and 0.05% Triton X-100. Long Term Storage: -80°C. Applications: ELISA Positive control.
Catalog No. 50-200-9650