Learn More
Drg International Inc DRG Chlamydia Trachomatis IgG ELISA RUO, 96 Well Kit
Supplier:  Drg International Inc EIA3462R
Encompass_Preferred
An enzyme immunoassay for the qualitative and semiquantitative determination of IgG-class antibodies to Chlamydia trachomatis in serum and plasma. This product is for Research Use Only. The DRG Chlamydia trachomatis IgG ELISA Kit is a solid phase enzyme-linked immunosorbent assay (ELISA). Microtiter wells as a solid phase are coated with inactivated Elementary Bodies Chlamydia trachomatis antigen. Diluted specimens and ready-for-use controls are pipetted into these wells.
Catalog No. 50-194-7563