Learn More
Medchemexpress LLC HY-P1237A 1mg , C-Type Natriuretic Peptide (CNP) (1-22), human (TFA) CAS:1966153-17-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP1237A1MG
Encompass_Preferred
HY-P1237A 1mg C-Type Natriuretic Peptide (CNP) (1-22), human (TFA) CAS: 1966153-17-2 C-Type Natriuretic Peptide (CNP) (1-22), human (TFA),a 1-22 fragment of CNP, is a natriuretic peptide receptor B (NPR-B) agonist. C-Type Natriuretic Peptide (CNP) (1-22), human (TFA) inhibits cAMP synthesis stimulated by the physiological agonists histamine and 5-HT or directly by Forskolin. CNP is a potent, endothelial-derived relaxant and growthinhibitory factor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1569