Learn More
Bachem C-Type Natriuretic Peptide (32-53) (human, porcine, rat) Acetate
SDP
Supplier:  Bachem 4095840.1000
Encompass_Preferred
C-Type Natriuretic Peptide (32-53) (human, porcine, rat) Acetate, H-Gly-Leu-Ser-Lys-Gly-Cys-Phe-Gly-Leu-Lys-Leu-Asp-Arg-Ile-Gly-Ser-Met-Ser-Gly-Leu-Gly-Cys-OH, 127869-51-6, C93H157N27O28S3, Mr 2197.63, 1mg. CNP-22 (human, porcine, rat), C-Type Natriuretic Peptide (32-53), human.
Catalog No. 50-194-6772