Learn More
Macherey-Nagel Syringe filters CHROMAFIL Xtra PTFE-20/25, BIGbox
Supplier:  Macherey-Nagel 729207.400
Encompass_Preferred
CHROMAFIL Xtra disposable syringe filters PTFE-20/25 BIGbox - membrane material: polytetrafluoroethylene - pore dia.: 0.2 µm, membrane dia.: 25 mm - colorless PP case, labeled - BIGbox of 400
Catalog No. NC0412451