Learn More
Elemental Scientific Inc C1-140228RH04-500 STANDARD
SDP
Catalog No. NC0656884
Encompass_Preferred
Custom Standard, APS WRF/PVNGS IC3000 High Anion B Primary, 500 ml 150 mg/L- PO4; 300m mg/L- F; 2000 mg/L- NO3 as N in H2O
Catalog No. NC0656884 Supplier Elemental Scientific Inc Supplier No. C1140228RH04500
Custom Standard, APS WRF/PVNGS IC3000 High Anion B Primary, 500 ml 150 mg/L- PO4; 300m mg/L- F; 2000 mg/L- NO3 as N in H2O