Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C1-C6 N-Paraffins in He, 1000 ppm Analytical Standard, MilliporeSigma™ Supelco™

Catalog No. 111022607 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
48 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-2607 48 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-2607 Supplier MilliporeSigma Supelco Supplier No. 501832

missing translation for 'validMainframeMsg'

Specifications

Packaging Cylinder
Flash Point Not Applicable
Physical Form Gas
UN Number UN 1956 2.2
Quantity 48 L
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.