Learn More
Medchemexpress LLC C1 Esterase Inhibito 500IU | 500IU
SDP
Catalog No. 5000324039
Encompass_Preferred
C1 Esterase Inhibitor (Human) is a C1 Esterase inhibitor derived from human plasma C1 Esterase Inhibitor (Human) a glycoprotein is a serum protease inhibitor (serpin) that binds covalently and inactivates C1r C1s and mannan-binding protein-associated proteases (MASPs) C1 Esterase Inhibitor (Human) has anti-inflammatory effects C1 Esterase Inhibitor (Human) can be used to prevent angioedema attacks associated with hereditary angioedema[1][2]
Catalog No. 50-003-24039 Supplier Medchemexpress LLC Supplier No. HYP991629500IU
Add to Cart
Add to Cart
C1 Esterase Inhibitor (Human) is a C1 Esterase inhibitor derived from human plasma C1 Esterase Inhibitor (Human) a glycoprotein is a serum protease inhibitor (serpin) that binds covalently and inactivates C1r C1s and mannan-binding protein-associated proteases (MASPs) C1 Esterase Inhibitor (Human) has anti-inflammatory effects C1 Esterase Inhibitor (Human) can be used to prevent angioedema attacks associated with hereditary angioedema[1][2]