Learn More
Athens Research And Technology Inc C1 Esterase Inhibitor, Native Human Plasma, ≥95% (SDS-PAGE), 1 mg
Catalog No. 50489676
Encompass_Preferred
C1 Esterase Inhibitor is a native protein purified from human plasma with ≥95% (SDS-PAGE), supplied frozen in a 1 mg pack. Molecular weight: 100 kDa. C-1 Esterase Inhibitor, Human Plasma: 1 MG; Frozen in 20 mM potassium phosphate, pH 7.0, with 250 mM potassium chloride; MW 100,000; EC=0.4; Purity: > =95% by SDS-PAGE. This protein is prepared and purified from human plasma. Common research applications include complement pathway, immunology and inflammation studies.
Catalog No. 50-489-676 Supplier Athens Research And Technology Inc Supplier No. 16160315091MG
C1 Esterase Inhibitor is a native protein purified from human plasma with ≥95% (SDS-PAGE), supplied frozen in a 1 mg pack. Molecular weight: 100 kDa. C-1 Esterase Inhibitor, Human Plasma: 1 MG; Frozen in 20 mM potassium phosphate, pH 7.0, with 250 mM potassium chloride; MW 100,000; EC=0.4; Purity: > =95% by SDS-PAGE. This protein is prepared and purified from human plasma. Common research applications include complement pathway, immunology and inflammation studies.