Learn More
Cayman Chemical C1 InhIbItr PolycLNal ntIb
SDP
Catalog No. 502917850
Encompass_Preferred
An isoflavonoid with antioxidant activity scavenges peroxynitrite nitric oxide radicals and total ROS (IC50s 1.36 1.13 and 6.51 UM respectively)
Catalog No. 50-291-7850 Supplier Cayman Chemical Supplier No. 38085100 UL
An isoflavonoid with antioxidant activity scavenges peroxynitrite nitric oxide radicals and total ROS (IC50s 1.36 1.13 and 6.51 UM respectively)