Learn More
Cayman Chemical C10 AV Ceramide (d18:1/10:0), Purity: ≥98%, CAS: 1263052-40-9, 25 mg
SDP
Catalog No. 502892428
Encompass_Preferred
A fluorescently tagged probe consisting of C-10 ceramide with an AV group attached to the end of the acyl chain
Catalog No. 50-289-2428 Supplier Cayman Chemical Supplier No. 900075325 MG
A fluorescently tagged probe consisting of C-10 ceramide with an AV group attached to the end of the acyl chain