Learn More
Medchemexpress LLC C10 Bisphosphonate | 15049-93-1 | ≥95.0% | C₁₀H₂₅NO₆P₂ | 1 MG
SDP
Catalog No. 5000359111
Encompass_Preferred
C10 Bisphosphonate is an acid sphingomyelinase (aSMase) inhibitor with an IC₅₀ of 0.02 μM in rats. It inhibits Dexamethasone-induced cell apoptosis and alleviates platelet-activating factor (PAF)-induced pulmonary edema. This compound can be used for the research of pulmonary edema.
  • Potently inhibits aSMase derived from crude rat brain extracts.
  • Effectively inhibits dexamethasone-induced apoptosis of HepG2 hepatocytes.
  • Reduces PAF-induced pulmonary edema in isolated perfused rats.
  • For research use only.
Catalog No. 50-003-59111 Supplier Medchemexpress LLC Supplier No. HY489171MG
C10 Bisphosphonate is an acid sphingomyelinase (aSMase) inhibitor with an IC₅₀ of 0.02 μM in rats. It inhibits Dexamethasone-induced cell apoptosis and alleviates platelet-activating factor (PAF)-induced pulmonary edema. This compound can be used for the research of pulmonary edema.
  • Potently inhibits aSMase derived from crude rat brain extracts.
  • Effectively inhibits dexamethasone-induced apoptosis of HepG2 hepatocytes.
  • Reduces PAF-induced pulmonary edema in isolated perfused rats.
  • For research use only.