Learn More
AdipoGen C10-DL-HSL
SDP
Supplier:  AdipoGen CDXD0338M010
Encompass_Preferred
Chemical. CAS 106983-36-2. Formula C14H25NO3. MW 255.4. Synthetic. N-Decanoyl-DL-homoserine lactone N-C10-HSL is a small diffusible signaling molecule and is a member of N-acyl-homoserine lactone family. N-acylhomoserine lactones AHL are involved in quorum sensing, controlling gene expression, and cellular metabolism. The diverse applications of this kind of molecule include regulation of virulence in general, infection prevention, and formation of biofilms.
Catalog No. 50-256-1104