Learn More
AdipoGen C10-L-HSL
SDP
Catalog No. 502561112
Encompass_Preferred
Chemical. CAS 177315-87-6. Formula C14H25NO3. MW 255.4. Synthetic. N-Decanoyl-L-homoserine lactone is a small diffusible signaling molecule involved in quorum sensing, thereby controlling gene expression and affecting cellular metabolism. The applications of this molecule include regulation of virulence and exoproteases.
Catalog No. 50-256-1112 Supplier AdipoGen Supplier No. CDXD0332M050
Chemical. CAS 177315-87-6. Formula C14H25NO3. MW 255.4. Synthetic. N-Decanoyl-L-homoserine lactone is a small diffusible signaling molecule involved in quorum sensing, thereby controlling gene expression and affecting cellular metabolism. The applications of this molecule include regulation of virulence and exoproteases.