Learn More
Bio Rad Laboratories C1000 Touch Thermal Cycler with 96–Deep Well Reaction Module
Catalog No. NC1705197
Encompass
Use our C1000 Touch thermal cycler with 96–deep well reaction module to run PCR reactions in plate or tube formats. Modular thermal cycler platform, includes C1000 Touch Thermal Cycler chassis, 96–deep well reaction module, USB flash drive.
This item is not returnable. View return policy
Catalog No. NC1705197 Supplier Bio Rad Laboratories Supplier No. 1851197
Add to Cart
Add to Cart
Request a Quote
Use our C1000 Touch thermal cycler with 96–deep well reaction module to run PCR reactions in plate or tube formats. Modular thermal cycler platform, includes C1000 Touch Thermal Cycler chassis, 96–deep well reaction module, USB flash drive.