Learn More
Aviva Systems Biology Corp C10orf12 Antibody - C-terminal region (ARP55319_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55319_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C10orf12. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human C10orf12. Alternate names: DKFZp564P1916, FLJ13022ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Pig, Rabbit, Rat
Catalog No. 50-286-028