Learn More
Aviva Systems Biology Corp C10orf132 Antibody - N-terminal region (ARP54405_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP54405_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C10orf132. It was validated on Western Blot. . Alternate names: C10orf132, C10orf133, MGC131701, bA451M19.3, bA459F3.4ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-285-923