Learn More
Aviva Systems Biology Corp C10orf30 Antibody - N-terminal region (ARP55523_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55523_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C10orf30. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human C10orf30. Alternate names: FLJ40283, MGC35247, C10orf30ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-286-049