Learn More
Aviva Systems Biology Corp C10orf33 Antibody - N-terminal region (ARP51276_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP51276_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C10orf33. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human C10orf33. Alternate names: FLJ23849, MGC13047, FP3420, C10orf33ApplicationWB. Species reactivity: Human
Catalog No. 50-014-60533