Learn More
Aviva Systems Biology Corp C10orf46 Antibody - middle region (ARP53203_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53203_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C10orf46. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human C10orf46. Alternate names: FLJ40409, MGC33215, C10orf46ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-60969