Learn More
Aviva Systems Biology Corp C10orf57 Antibody - middle region (ARP49927_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49927_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C10orf57. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human C10orf57. Alternate names: FLJ13263, bA369J21.6, C10orf57, RP11-369J21.6ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-014-60217