Learn More
Aviva Systems Biology Corp C10orf96 Antibody - middle region (ARP55892_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55892_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C10orf96. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human C10orf96. Alternate names: MGC35062, C10orf96ApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Pig, Rabbit, Rat
Catalog No. 50-014-61651