Learn More
AdipoGen N-Undecanoyl-L-homoserine lactone (10 mg)
SDP
Supplier:  AdipoGen CDXU0026M010
Encompass_Preferred
Chemical. CAS: 216596-71-3. Formula: C15H27NO3. MW: 269.39. N-Undecanoyl-L-homoserine lactone lactone is a small diffusible signaling molecule and is a member of N-acyl-homoserine lactone family. N-acylhomoserine lactones (AHL) are involved in quorum sensing, controlling gene expression, and cellular metabolism. The diverse applications of this kind of molecule include regulation of virulence in general, infection prevention, and formation of biofilms. It appears later than shorter acyl chain AHLs in developing biofilms and, like other long chain AHLs, stimulates bacterial growth. C11-HSL possesses a rare odd-numbered acyl carbon chain and may be a minor quorum-sensing signaling molecule in P. aeruginosa strains.
Catalog No. 50-205-4678