Learn More
Aviva Systems Biology Corp C11orf24 Antibody - N-terminal region (ARP49708_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49708_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C11orf24. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human C11orf24. Alternate names: DM4E3ApplicationIHC, WB. Species reactivity: Human
Catalog No. 50-014-60152