Learn More
Aviva Systems Biology Corp C11orf42 Antibody - C-terminal region (ARP53268_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53268_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C11orf42. It was validated on Western Blot. . Alternate names: MGC34805ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-820