Learn More
Aviva Systems Biology Corp C11orf53 Antibody - middle region (ARP55885_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55885_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C11orf53. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human C11orf53. Alternate names: MGC50104ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-61648