Learn More
Aviva Systems Biology Corp C11orf54 Antibody - C-terminal region (ARP41594_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41594_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C11orf54. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human C11orf54. Alternate names: PTD012ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-53308