Learn More
Aviva Systems Biology Corp C11orf74 Antibody - middle region (ARP52634_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52634_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C11orf74. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human C11orf74. Alternate names: FLJ38678ApplicationWB. Species reactivity: Human
Catalog No. 50-014-60789