Learn More
Medchemexpress LLC C12-200 | 1220890-25-4 | MFCD32062847 | 99.9% | C70H145N5O5 | 10MG
SDP
Catalog No. 5000259387
Encompass_Preferred
C12-200 is an ionizable cationic lipidoid used as a helper lipid in lipid nanoparticle (LNP) formulations to enable efficient delivery of nucleic acids such as mRNA and siRNA. It supports encapsulation, cellular uptake, and endosomal release, and is intended for formulation research in vitro and in vivo.
  • Ionizable cationic lipid for mRNA and siRNA delivery.
  • Facilitates efficient encapsulation into lipid nanoparticles.
  • Promotes endosomal escape and improved intracellular delivery.
  • High purity suitable for formulation research (99.9%).
  • Available in small research pack sizes for laboratory use.
Catalog No. 50-002-59387 Supplier Medchemexpress LLC Supplier No. HY14540510MG
C12-200 is an ionizable cationic lipidoid used as a helper lipid in lipid nanoparticle (LNP) formulations to enable efficient delivery of nucleic acids such as mRNA and siRNA. It supports encapsulation, cellular uptake, and endosomal release, and is intended for formulation research in vitro and in vivo.
  • Ionizable cationic lipid for mRNA and siRNA delivery.
  • Facilitates efficient encapsulation into lipid nanoparticles.
  • Promotes endosomal escape and improved intracellular delivery.
  • High purity suitable for formulation research (99.9%).
  • Available in small research pack sizes for laboratory use.