Learn More
Broadpharm C12-200, 250 mg, Liquid, Branched-chain ionizable cationic lipidoid, Soluble in Ethanol, DMSO, DMF, CAS 1220890-25-4
Catalog No. NC2170156
Encompass
C12-200 is a branched-chain ionizable cationic lipidoid developed for efficient delivery in lipid nanoparticle (LNP) formulations. This product features five hydroxyl groups which allow for enhanced functionalization, making it suitable for intracellular delivery of therapeutic agents such as mRNA and siRNA. With significant improvements in formulation potency, including a notable 7-fold increase for erythropoietin-mRNA-loaded nanoparticles, C12-200 is essential for advancements in vaccine development and gene editing.
  • Liquid form for ease of use
  • Designed for lipid nanoparticle (LNP) formation
  • Soluble in ethanol, DMSO, and DMF
  • Stores at -20°C; protect from light
  • Facilitates delivery of mRNA, siRNA, and proteins
  • Provides opportunities for further chemical derivatization
  • Significantly increases formulation potency
This item is not returnable. View return policy
Catalog No. NC2170156 Supplier Broadpharm Supplier No. BP26474250MG
C12-200 is a branched-chain ionizable cationic lipidoid developed for efficient delivery in lipid nanoparticle (LNP) formulations. This product features five hydroxyl groups which allow for enhanced functionalization, making it suitable for intracellular delivery of therapeutic agents such as mRNA and siRNA. With significant improvements in formulation potency, including a notable 7-fold increase for erythropoietin-mRNA-loaded nanoparticles, C12-200 is essential for advancements in vaccine development and gene editing.
  • Liquid form for ease of use
  • Designed for lipid nanoparticle (LNP) formation
  • Soluble in ethanol, DMSO, and DMF
  • Stores at -20°C; protect from light
  • Facilitates delivery of mRNA, siRNA, and proteins
  • Provides opportunities for further chemical derivatization
  • Significantly increases formulation potency