Learn More
AdipoGen C12-DL-HSL
SDP
Supplier:  AdipoGen CDXD0339M010
Encompass_Preferred
Chemical. CAS 18627-38-8. Formula C16H29NO3. MW 283.4. Synthetic. N-Dodecanoyl-DL-homoserine lactone C12-HSL is a small diffusible signalling molecule involved in quorum sensing, thereby controlling gene expression and affecting cellular metabolism in bacteria. In addition to regulating bacterial functions, C12-HSL activates NF-kappaB in RAW 264.7 macrophages, increasing the expression of TNF-alpha, interleukin-1beta IL-1beta and IL-8, while other lactones do not. In addition, C12-HSL alters cell cycling and metabolism of human keratinocyte HaCaT cells. It also was shown to induce cell death by apoptosis.
Catalog No. 50-256-1102