Learn More
Cayman Chemical C12 Galactosylceramide (d18:1/12:0), 41613-14-3, 1 mg
SDP
Supplier:  Cayman Chemical 243471
Encompass_Preferred
A bioactive sphingolipid; inhibits IL-4 production by 53.84% in EL4 T cells (10 μM); reduces the growth of human papillomavirus type 16-associated tumors in mice; reduces natural killer T cell activity, delays the onset of proteinuria, and improves survival in a mouse model of systemic lupus
Catalog No. NC1472684