Learn More
Cayman Chemical C12 Galactosylceramide (d18:1/12:0), Purity: ≥95%, CAS: 41613-14-3, 50 mg
SDP
Catalog No. 502934857
Encompass_Preferred
A bioactive sphingolipid; inhibits IL-4 production by 53.84% in EL4 T cells (10 μM); reduces the growth of human papillomavirus type 16-associated tumors in mice; reduces natural killer T cell activity, delays the onset of proteinuria, and improves survival in a mouse model of systemic lupus
Catalog No. 50-293-4857 Supplier Cayman Chemical Supplier No. 2434750 MG
A bioactive sphingolipid; inhibits IL-4 production by 53.84% in EL4 T cells (10 μM); reduces the growth of human papillomavirus type 16-associated tumors in mice; reduces natural killer T cell activity, delays the onset of proteinuria, and improves survival in a mouse model of systemic lupus