Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC C12-iE-DAP hydrochloride | 1269619-57-9 | 538.07 | 10 MG
SDP

Catalog No. 5000398026
Encompass_Preferred

C12-iE-DAP hydrochloride is the hydrochloride salt form of C12-iE-DAP, a biologically active peptide that acts as an agonist for NOD1. It is designed for high-speed applications and is suitable for laboratory centrifugation and storage.

  • Molecular formula: C24H44ClN3O8
  • Sequence: Lauroyl-γ-d-Glu-meso-diaminopimelic acid
  • Target: NOD-like Receptor (NLR)
  • Pathway: Immunology/Inflammation
  • Solvent & solubility (in vitro): DMSO: 100 mg/mL (185.85 mM; need ultrasonic)
  • Agonistic effects on NOD1

Catalog No. 50-003-98026 Supplier Medchemexpress LLC Supplier No. HYP5524A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

C12-iE-DAP hydrochloride is the hydrochloride salt form of C12-iE-DAP, a biologically active peptide that acts as an agonist for NOD1. It is designed for high-speed applications and is suitable for laboratory centrifugation and storage.

  • Molecular formula: C24H44ClN3O8
  • Sequence: Lauroyl-γ-d-Glu-meso-diaminopimelic acid
  • Target: NOD-like Receptor (NLR)
  • Pathway: Immunology/Inflammation
  • Solvent & solubility (in vitro): DMSO: 100 mg/mL (185.85 mM; need ultrasonic)
  • Agonistic effects on NOD1

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.