Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC C12-iE-DAP hydrochloride | 538.07 | 1 MG
SDP

Catalog No. 5000399095
Encompass_Preferred

C12-iE-DAP hydrochloride is the hydrochloride salt form of C12-iE-DAP, a biologically active peptide with agonistic effects on NOD1. It targets the NOD-like Receptor (NLR) and plays a role in immunology and inflammation pathways.

  • Hydrochloride salt form
  • Biologically active peptide
  • Agonistic effects on NOD1
  • Molecular formula: C24H44ClN3O8
  • Sequence: Lauroyl-γ-d-Glu-meso-diaminopimelic acid
  • Targets NOD-like Receptor (NLR)
  • Involved in immunology/inflammation pathways

Catalog No. 50-003-99095 Supplier Medchemexpress LLC Supplier No. HYP5524A1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

C12-iE-DAP hydrochloride is the hydrochloride salt form of C12-iE-DAP, a biologically active peptide with agonistic effects on NOD1. It targets the NOD-like Receptor (NLR) and plays a role in immunology and inflammation pathways.

  • Hydrochloride salt form
  • Biologically active peptide
  • Agonistic effects on NOD1
  • Molecular formula: C24H44ClN3O8
  • Sequence: Lauroyl-γ-d-Glu-meso-diaminopimelic acid
  • Targets NOD-like Receptor (NLR)
  • Involved in immunology/inflammation pathways

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.