Learn More
Cayman Chemical C12 Lissamine-rhodamine Galac
SDP
Catalog No. 502935731
Encompass_Preferred
A fluorescently labeled version of C12 galactosylceramide; ex/em = 560/575 nm
Catalog No. 50-293-5731 Supplier Cayman Chemical Supplier No. 24470500 UG
A fluorescently labeled version of C12 galactosylceramide; ex/em = 560/575 nm