Learn More
Cayman Chemical C12 NBD dihydro Ceramide d18
SDP
Catalog No. 502889990
Encompass_Preferred
A fluorescent ceramide analog that contains a saturated bond in the C-4/C-5 position of the sphingosine backbone
Catalog No. 50-288-9990 Supplier Cayman Chemical Supplier No. 10008097500 UG
A fluorescent ceramide analog that contains a saturated bond in the C-4/C-5 position of the sphingosine backbone