Learn More
Cayman Chemical C12 NBD dihydro Ceramide (d18:0/12:0), Purity: ≥98%, CAS: 474943-05-0, 5 mg
SDP
Catalog No. 502887729
Encompass_Preferred
A fluorescent ceramide analog that contains a saturated bond in the C-4/C-5 position of the sphingosine backbone
Catalog No. 50-288-7729 Supplier Cayman Chemical Supplier No. 100080975 MG
A fluorescent ceramide analog that contains a saturated bond in the C-4/C-5 position of the sphingosine backbone