Learn More
Cayman Chemical C12 NBD GlobotrIaosylcRamID
SDP
Catalog No. 502935748
Encompass_Preferred
A fluorescently tagged sphingolipid; a derivative of globotriaosylceramide that is tagged with a fluorescent C12 NBD; accumulates in HUVECs treated with DGJ in a concentration-dependent manner
Catalog No. 50-293-5748 Supplier Cayman Chemical Supplier No. 24628100 UG
A fluorescently tagged sphingolipid; a derivative of globotriaosylceramide that is tagged with a fluorescent C12 NBD; accumulates in HUVECs treated with DGJ in a concentration-dependent manner