Learn More
Cayman Chemical C12 NBD PhytocRamIDt18 0 100ug
SDP
Catalog No. 502936397
Encompass_Preferred
A fluorescently tagged derivative of phytosphingosine that has been used to study the metabolism of ceramides
Catalog No. 50-293-6397 Supplier Cayman Chemical Supplier No. 24327100 UG
A fluorescently tagged derivative of phytosphingosine that has been used to study the metabolism of ceramides