Learn More
Medchemexpress LLC C12-TLRa | 3067466-15-0 | 99.0% | 728.10 g/mol | C46H73N5O2 | 10 MG
SDP
Catalog No. 5000287169
Encompass_Preferred
C12-TLRa is an ionized adjuvant lipid that acts as a toll-like receptor 7/8 (TLR7/8) agonist in endosomes. It is used as a structural component in lipid nanoparticles to enhance mRNA delivery and to stimulate innate immune responses; common research uses include mRNA vaccine formulation and immunostimulatory studies.
  • Ionized adjuvant lipid targeting TLR7/8 receptors.
  • Enhances mRNA delivery when formulated in lipid nanoparticles.
  • Promotes innate immune responses for vaccine research.
  • High purity: 99.0%.
  • Molecular weight 728.10 g/mol; formula C46H73N5O2.
  • Provided as a solid suitable for formulation and analytical use.
Catalog No. 50-002-87169 Supplier Medchemexpress LLC Supplier No. HY15387910MG
C12-TLRa is an ionized adjuvant lipid that acts as a toll-like receptor 7/8 (TLR7/8) agonist in endosomes. It is used as a structural component in lipid nanoparticles to enhance mRNA delivery and to stimulate innate immune responses; common research uses include mRNA vaccine formulation and immunostimulatory studies.
  • Ionized adjuvant lipid targeting TLR7/8 receptors.
  • Enhances mRNA delivery when formulated in lipid nanoparticles.
  • Promotes innate immune responses for vaccine research.
  • High purity: 99.0%.
  • Molecular weight 728.10 g/mol; formula C46H73N5O2.
  • Provided as a solid suitable for formulation and analytical use.