Learn More
Medchemexpress LLC C12-TLRa | 3067466-15-0 | C46H73N5O2 | 5 MG
SDP
Catalog No. 5000376253
Encompass_Preferred
C12-TLRa is an ionized adjuvant lipid that targets TLR7/8 receptors in endosomes. It enhances mRNA delivery as a structural component of LNPs and boosts innate immune responses. This compound can be utilized for research related to SARS-CoV-2 mRNA vaccines.
  • Ionized adjuvant lipid
  • Targets TLR7/8 receptors in endosomes
  • Enhances messenger RNA delivery as a structural component of lipid nanoparticles
  • Enhances innate immune responses
  • Potential use in SARS-CoV-2 messenger RNA vaccine research
Catalog No. 50-003-76253 Supplier Medchemexpress LLC Supplier No. HY1538795MG
C12-TLRa is an ionized adjuvant lipid that targets TLR7/8 receptors in endosomes. It enhances mRNA delivery as a structural component of LNPs and boosts innate immune responses. This compound can be utilized for research related to SARS-CoV-2 mRNA vaccines.
  • Ionized adjuvant lipid
  • Targets TLR7/8 receptors in endosomes
  • Enhances messenger RNA delivery as a structural component of lipid nanoparticles
  • Enhances innate immune responses
  • Potential use in SARS-CoV-2 messenger RNA vaccine research