Learn More
Bachem Dodecyl pentaethylene glycolether
SDP
Supplier:  Bachem 4028288.0001
Encompass_Preferred
Dodecyl pentaethylene glycolether, 3055-95-6, C22H46O6, Mr 406.60, 1g. Pentaethyleneglycolmonododecylether, C12E5. C12E5, nonionic surfactant useful for the extraction of membrane proteins, for example the fusion protein and the haemagglutininneuraminidase of the Sendai virus.
Catalog No. 50-194-7412