Learn More
Aviva Systems Biology Corp INTS13 Antibody - middle region (ARP57133_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57133_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C12orf11. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human C12orf11. Alternate names: FLJ10630, FLJ10637, NET48, Mat89Bb, C12orf11ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-54014