Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC C13H9N5O5S2 | 883065-90-5 | MFCD01313575 | 99.9% | 379.37 g/mol | C13H9N5O5S2 | 100 MG
SDP

Catalog No. 5000245096
Encompass_Preferred

BI-78D3 is a small-molecule substrate-competitive inhibitor of c-Jun N-terminal kinases (JNK) used as a research tool to probe JNK-dependent signaling. It inhibits JNK kinase activity with a reported IC50 of 280 nM and is supplied for in vitro research use only.

  • Substrate-competitive JNK inhibitor (IC50 = 280 nM).
  • Molecular formula C13H9N5O5S2; molecular weight 379.37 g/mol.
  • High purity (99.85%) suitable for biochemical and cell-based assays.
  • Soluble in DMSO; handle using standard laboratory safety procedures.
  • For research use only; not for human or veterinary applications.

Catalog No. 50-002-45096 Supplier Medchemexpress LLC Supplier No. HY10366100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

BI-78D3 is a small-molecule substrate-competitive inhibitor of c-Jun N-terminal kinases (JNK) used as a research tool to probe JNK-dependent signaling. It inhibits JNK kinase activity with a reported IC50 of 280 nM and is supplied for in vitro research use only.

  • Substrate-competitive JNK inhibitor (IC50 = 280 nM).
  • Molecular formula C13H9N5O5S2; molecular weight 379.37 g/mol.
  • High purity (99.85%) suitable for biochemical and cell-based assays.
  • Soluble in DMSO; handle using standard laboratory safety procedures.
  • For research use only; not for human or veterinary applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.