Learn More
Aviva Systems Biology Corp C13orf28 Antibody - middle region (ARP48009_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48009_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C13orf28. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human C13orf28. Alternate names: FLJ27356, C13orf28ApplicationWB. Species reactivity: Human
Catalog No. 50-014-54011