Learn More
Aviva Systems Biology Corp C13orf31 Antibody - middle region (ARP53139_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53139_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C13orf31. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human C13orf31. Alternate names: DKFZp686D11119, FLJ38725, C13orf31, RP11-5G9.2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-60940