Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC C14-4 | 2639634-80-1 | 99.0% | 1365.30 g/mol | C84H173N5O7 | 10 MG
SDP

Catalog No. 5000285388
Encompass_Preferred

C14-4 is an ionizable lipid used to formulate lipid nanoparticles (LNPs) for mRNA delivery and related research applications. Supplied as a liquid with high reported purity, it is soluble in common organic solvents and suitable for both in vitro and in vivo formulation workflows.

  • Ionizable lipid for lipid nanoparticle formulation.
  • Enhances mRNA delivery and cellular uptake.
  • High purity (≈99.0%).
  • Soluble in DMSO and ethanol at 100 mg/mL.
  • Liquid appearance, colorless to light yellow.
  • Storage: pure form -20°C; in solvent -80°C for 6 months or -20°C for 1 month.
  • Formulation compatible: clear solution in 10% DMSO/90% corn oil at ≥2.5 mg/mL.

Catalog No. 50-002-85388 Supplier Medchemexpress LLC Supplier No. HY14884210MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

C14-4 is an ionizable lipid used to formulate lipid nanoparticles (LNPs) for mRNA delivery and related research applications. Supplied as a liquid with high reported purity, it is soluble in common organic solvents and suitable for both in vitro and in vivo formulation workflows.

  • Ionizable lipid for lipid nanoparticle formulation.
  • Enhances mRNA delivery and cellular uptake.
  • High purity (≈99.0%).
  • Soluble in DMSO and ethanol at 100 mg/mL.
  • Liquid appearance, colorless to light yellow.
  • Storage: pure form -20°C; in solvent -80°C for 6 months or -20°C for 1 month.
  • Formulation compatible: clear solution in 10% DMSO/90% corn oil at ≥2.5 mg/mL.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.