Learn More
TARGETMOL CHEMICALS INC C14 Ceramide 10MG
SDP
Catalog No. 502876689
Encompass_Preferred
Also available in 1 mg, 5 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. C14 Ceramide is an endogenous ceramide produced by ceramide synthetase 6, which can be used for Parkinson's research. Purity 99.63%
Catalog No. 50-287-6689 Supplier TARGETMOL CHEMICALS INC Supplier No. T3580310MG
Also available in 1 mg, 5 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. C14 Ceramide is an endogenous ceramide produced by ceramide synthetase 6, which can be used for Parkinson's research. Purity 99.63%